accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3NH2O

CAS RN 56108-12-4

IUPAC Name: 4-tert-butylbenzamide
Synonyms: 4-tert-butylbenzamide
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: VIPMBJSGYWWHAO-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results