missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzamide, 98%

Product Code. 11458326
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458326 50 g 50g
11448326 10 g 10g
11468326 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458326 Supplier Thermo Scientific Alfa Aesar Supplier No. A15380.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56108-12-4 | C11H15NO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56108-12-4
Molecular Formula C11H15NO
MDL Number MFCD00017139
Synonym 4-tert-butyl benzamide, benzamide, 4-1,1-dimethylethyl, p-tert-butyl benzamide, 4-tert-butyl benzenecarboxamide, unii-5e7907z6bm, 4-1,1-dimethylethyl benzamide, p-t-butylbenzamide, 4-t-butylbenzamide, acmc-20amap, butylbenzenecarboxamide

Specifications

Melting Point 171°C to 175°C
Quantity 50 g
Beilstein 2086692
Formula Weight 177.25
Percent Purity 98%
Chemical Name or Material 4-tert-Butylbenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.