accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5614-37-9

IUPAC Name: methoxycyclopentane
Synonyms: cyclopentyl methyl ether
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: SKTCDJAMAYNROS-UHFFFAOYSA-N
SMILES: COC1CCCC1
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (4)
Grade 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (2)
  • (4)
  • (2)
  • (4)
  • (6)