CAS RN 565453-39-6
CAS RN 565453-39-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Vildagliptin Amide, TRC
CAS: 565453-39-6 Synonym: N-(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)glycyl-L-prolinamide∣Vildagliptin Amide∣Vildagliptin Impurity A