missing translation for 'onlineSavingsMsg'
Learn More

Vildagliptin Amide, TRC

Product Code. 17990445
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17990445 100 mg 100mg
17943955 50 mg 50mg
17981195 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17990445 Supplier Toronto Research Chemicals Supplier No. TRCH971380100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565453-39-6
Synonym N-(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)glycyl-L-prolinamide∣Vildagliptin Amide∣Vildagliptin Impurity A

Specifications

Purity >95% (HPLC)
Format Neat
Molecular Formula C17H27N3O3
MDL Number MFCD30541494
Molecular Weight (g/mol) 321.41, 321.42
InChI Key IUJJPEBUSBVYIP-VHDAAVTISA-N
SMILES [H][C@]12C[C@]3([H])CC(O)(C1)CC(C2)(C3)NCC(=O)N1CCCC1C(N)=O
IUPAC Name 1-(2-{[(5R,7S)-3-hydroxyadamantan-1-yl]amino}acetyl)pyrrolidine-2-carboxamide
Product Type Metabolite
Quantity 100 mg
Shipping Condition Room Temperature
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.