CAS RN 566-99-4
CAS RN 566-99-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Doristerol, TRC
CAS: 566-99-4 Molecular Formula: C27H46O Molecular Weight (g/mol): 386.65 Synonym: (3beta,5alpha)-Cholest-8(14)-en-3-ol,5alpha-Cholest-8(14)-ene-3beta-ol SMILES: CC(C)CCC[C@@H](C)[C@H]1CCC2=C3CC[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C