missing translation for 'onlineSavingsMsg'
Learn More

Doristerol, TRC

Product Code. 30387268
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387268 50 mg
30375611 100 mg
30386300 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387268 Supplier Toronto Research Chemicals Supplier No. TRCD53178050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 566-99-4
Molecular Formula C27H46O
Molecular Weight (g/mol) 386.65
Synonym (3beta,5alpha)-Cholest-8(14)-en-3-ol, 5alpha-Cholest-8(14)-ene-3beta-ol
SMILES CC(C)CCC[C@@H](C)[C@H]1CCC2=C3CC[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h18-21,23,25,28H,6-17H2,1-5H3/t19-,20+,21+,23-,25+,26+,27-/m1/s1
Formula Weight 386.3549
Chemical Name or Material Doristerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.