accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONHNO

CAS RN 56795-92-7

IUPAC Name: 5-(benzyloxy)-1H-pyrrolo[2,3-c]pyridine-3-carbaldehyde
Synonyms: 5-(benzyloxy)-1H-pyrrolo[2,3-c]pyridine-3-carbaldehyde
Molecular Weight (g/mol): 252.27
Molecular Formula: C15H12N2O2
InChi Key: OZWLZROTVDCROC-UHFFFAOYSA-N
SMILES: O=CC1=CNC2=CN=C(OCC3=CC=CC=C3)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results