accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 569-64-2

ClH3CNCH3CNCH3H3C

CAS RN 569-64-2

IUPAC Name: bis[4-(dimethylamino)phenyl](phenyl)methylium chloride
Synonyms: malachite green
Molecular Weight (g/mol): 364.92
Molecular Formula: C23H25ClN2
InChi Key: FDZZZRQASAIRJF-UHFFFAOYSA-M
SMILES: [Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results