CAS RN 57-48-7
D(-)-Fructose, specified according the requirements of USP
CAS: 57-48-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.16 InChI Key: BJHIKXHVCXFQLS-UYFOZJQFSA-N Synonym: d---fructose,d--fructose,3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one,furucton,arabino-hexulose,keto-d-fructose,d-levulose,sugar, fruit,fructose, d,krystar 300 PubChem CID: 5984 ChEBI: CHEBI:48095 IUPAC Name: (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one SMILES: C(C(C(C(C(=O)CO)O)O)O)O
Thermo Scientific Chemicals D(-)-Fructose, 99%
CAS: 57-48-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.16 InChI Key: BJHIKXHVCXFQLS-UYFOZJQFSA-N Synonym: d---fructose,d--fructose,3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one,furucton,arabino-hexulose,keto-d-fructose,d-levulose,sugar, fruit,fructose, d,krystar 300 PubChem CID: 5984 ChEBI: CHEBI:48095 IUPAC Name: (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one SMILES: C(C(C(C(C(=O)CO)O)O)O)O
Thermo Scientific Chemicals D-Fructose, 99%
CAS: 57-48-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 MDL Number: MFCD00148910 InChI Key: BJHIKXHVCXFQLS-UYFOZJQFSA-N Synonym: d---fructose,d--fructose,3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one,furucton,arabino-hexulose,keto-d-fructose,d-levulose,sugar, fruit,fructose, d,krystar 300 PubChem CID: 5984 ChEBI: CHEBI:48095 IUPAC Name: (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one SMILES: C(C(C(C(C(=O)CO)O)O)O)O
D(-)-Fructose, Certified AR for Analysis, Fisher Chemical™
CAS: 57-48-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 MDL Number: 148910 InChI Key: BJHIKXHVCXFQLS-UYFOZJQFSA-N Synonym: d---fructose,d--fructose,3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one,furucton,arabino-hexulose,keto-d-fructose,d-levulose,sugar, fruit,fructose, d,krystar 300 PubChem CID: 5984 ChEBI: CHEBI:48095 IUPAC Name: (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one SMILES: C(C(C(C(C(=O)CO)O)O)O)O
D-Fructose, TRC
CAS: 57-48-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.16 Synonym: Fructose,(-)-d-Arabino-hex-2-ulopyranose,D(-)-Fructose IUPAC Name: (2RS,3S,4R,5R)-2-(hydroxymethyl)oxane-2,3,4,5-tetrol SMILES: O[C@]1(CO)O[C@H](CO)[C@@H](O)[C@@H]1O
Fructose, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Melford D-Fructose
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
D(-)-Fructose, Pure, Fisher Chemical™
CAS: 57-48-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 MDL Number: 148910 InChI Key: BJHIKXHVCXFQLS-UYFOZJQFSA-N Synonym: d---fructose,d--fructose,3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one,furucton,arabino-hexulose,keto-d-fructose,d-levulose,sugar, fruit,fructose, d,krystar 300 PubChem CID: 5984 ChEBI: CHEBI:48095 IUPAC Name: (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one SMILES: C(C(C(C(C(=O)CO)O)O)O)O
Fructose, LoGiCal
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.