accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3(R)H3C(R)CH3(R)(R)(S)OH(R)H3C(S)(R)H3C(E)

CAS RN 57-87-4

IUPAC Name: (1R,3aR,7S,9aR,9bS,11aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Synonyms: ergosterol
Molecular Weight (g/mol): 396.66
Molecular Formula: C28H44O
InChi Key: DNVPQKQSNYMLRS-APGDWVJJSA-N
SMILES: CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2C3=CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Molecular Weight (g/mol) 
  • (3)
  • (6)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
Certifications/Compliance 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)