accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHN

CAS RN 574-66-3

IUPAC Name: N-(diphenylmethylidene)hydroxylamine
Synonyms: methanone, diphenyl-, oxime
Molecular Weight (g/mol): 197.24
Molecular Formula: C13H11NO
InChi Key: DNYZBFWKVMKMRM-UHFFFAOYSA-N
SMILES: ON=C(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results