accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CO

CAS RN 57402-36-5

IUPAC Name: docosanal
Synonyms: behenoyl
Molecular Weight (g/mol): 324.59
Molecular Formula: C22H44O
InChi Key: ULCXRAFXRZTNRO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCC=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)