CAS RN 57469-77-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ibuprofen Lysinate, TRC
CAS: 57469-77-9 Molecular Formula: C13 H18 O2 . C6 H14 N2 O2 Molecular Weight (g/mol): 352.468 Synonym: L-Lysine, α-methyl-4-(2-methylpropyl)benzeneacetate (1:1) (ACI),L-Lysine, mono[α-methyl-4-(2-methylpropyl)benzeneacetate] (9CI),Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, compd. with L-lysine (1:1) (9CI),Arfen,Dolormin,Ibuprofen lysinate,Ibuprofen lysine,Ibuprofen lysine salt,Imbun,Imbun 500,Lisiprofen,Lysine 2-(p-isobutylphenyl)propionate,NeoProfen,Saren,Solufenum,Soluphene IUPAC Name: (2S)-2,6-diaminohexanoic acid;2-[4-(2-methylpropyl)phenyl]propanoic acid SMILES: CC(C)Cc1ccc(cc1)C(C)C(=O)O.NCCCC[C@H](N)C(=O)O