missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen Lysinate, TRC

Product Code. 30133463
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133463 10 g 10g
30041128 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133463 Supplier Toronto Research Chemicals Supplier No. TRCI14009510G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57469-77-9
Molecular Formula C13 H18 O2 . C6 H14 N2 O2
Molecular Weight (g/mol) 352.468
Synonym L-Lysine, α-methyl-4-(2-methylpropyl)benzeneacetate (1:1) (ACI), L-Lysine, mono[α-methyl-4-(2-methylpropyl)benzeneacetate] (9CI), Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, compd. with L-lysine (1:1) (9CI), Arfen, Dolormin, Ibuprofen lysinate, Ibuprofen lysine, Ibuprofen lysine salt, Imbun, Imbun 500, Lisiprofen, Lysine 2-(p-isobutylphenyl)propionate, NeoProfen, Saren, Solufenum, Soluphene
IUPAC Name (2S)-2,6-diaminohexanoic acid;2-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)Cc1ccc(cc1)C(C)C(=O)O.NCCCC[C@H](N)C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H18O2.C6H14N2O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15;7-4-2-1-3-5(8)6(9)10/h4-7,9-10H,8H2,1-3H3,(H,14,15);5H,1-4,7-8H2,(H,9,10)/t;5-/m.0/s1
Formula Weight 352.236
Percent Purity >95%
Chemical Name or Material (2S)-2,6-diaminohexanoic acid 2-[4-(2-methylpropyl)phenyl]propanoic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.