accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 577-16-2

IUPAC Name: 1-(2-methylphenyl)ethan-1-one
Synonyms: ethanone, 1-(2-methylphenyl)-
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: YXWWHNCQZBVZPV-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC=C1C
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results