accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOHO

CAS RN 577-56-0

IUPAC Name: 2-acetylbenzoic acid
Synonyms: O-acetylbenzoic acid
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: QDAWXRKTSATEOP-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC=C1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)