accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClBr

CAS RN 578-51-8

IUPAC Name: 1-bromo-2-(chloromethyl)benzene
Synonyms: 1-bromo-2-(chloromethyl)benzene
Molecular Weight (g/mol): 205.48
Molecular Formula: C7H6BrCl
InChi Key: DDVSFIUKWUTKES-UHFFFAOYSA-N
SMILES: ClCC1=CC=CC=C1Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results