Learn More
2-Bromobenzyl chloride, 97%
Product Code. 11318525
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318525 5 g 5g
11308525 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11318525 Supplier Thermo Scientific Alfa Aesar Supplier No. L11953.06
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 578-51-8 | C7H6BrCl | 205.479 g/mol
2-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 578-51-8
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.479
MDL Number MFCD00040863
InChI Key DDVSFIUKWUTKES-UHFFFAOYSA-N
Synonym 1-bromo-2-chloromethyl benzene, 2-bromobenzyl chloride, o-bromobenzyl chloride, benzene,1-bromo-2-chloromethyl, benzene, 1-bromo-2-chloromethyl, o-brombenzylchlorid, acmc-1aunq, 3-bromo-2-chloromethylbenzene, 1-bromo-2-chloromethyl benzene #
PubChem CID 521816
IUPAC Name 1-bromo-2-(chloromethyl)benzene
SMILES C1=CC=C(C(=C1)CCl)Br
Specifications
Boiling Point 110°C to 111°C (15 mmHg)
Refractive Index 1.586
Quantity 5 g
UN Number UN3265
Beilstein 471701
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 205.49
Percent Purity 97%
Chemical Name or Material 2-Bromobenzyl chloride