accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONBr

CAS RN 57883-25-7

IUPAC Name: 3-bromo-2-ethoxypyridine
Synonyms: 3-bromo-2-ethoxypyridine
Molecular Weight (g/mol): 202.05
Molecular Formula: C7H8BrNO
InChi Key: KLQBFCAXXZMOLZ-UHFFFAOYSA-N
SMILES: CCOC1=C(Br)C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results