accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(S)(S)(R)O(R)H3C(S)(S)HO

CAS RN 58-22-0

IUPAC Name: (1S,3aS,3bR,9aR,9bS,11aS)-1-hydroxy-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: testosterone β testosterone andro andriol android 5 androgel AndroGel testim android 10 total testosterone
Molecular Weight (g/mol): 288.43
Molecular Formula: C19H28O2
InChi Key: MUMGGOZAMZWBJJ-DYKIIFRCSA-N
SMILES: C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O
Molecular Weight (g/mol) 
  • (10)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Certifications/Compliance 
  • (3)
  • (1)
Industry Type 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)
  • (10)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)