accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH2COOOHOClCl

CAS RN 58-54-8

IUPAC Name: 2-[2,3-dichloro-4-(2-methylidenebutanoyl)phenoxy]acetic acid
Synonyms: ethacrynic acid ethacrynic acid (substance) hidromedin uregit methylenebutyryl phenoxyacetic acid ethacrynate sodium preparation sodium, ethacrynate ethacrynic acid, sodium salt methylenebutyrylphenoxyacetic acid acido etacrinico
Molecular Weight (g/mol): 303.14
Molecular Formula: C13H12Cl2O4
InChi Key: AVOLMBLBETYQHX-UHFFFAOYSA-N
SMILES: CCC(=C)C(=O)C1=CC=C(OCC(O)=O)C(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)