accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 581-89-5

ONO

CAS RN 581-89-5

IUPAC Name: 2-nitronaphthalene
Synonyms: 2-nitronaphthalene
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: ZJYJZEAJZXVAMF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)