missing translation for 'onlineSavingsMsg'
Learn More

2-Nitronaphthalene, TRC

Product Code. 30011271
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011271 1 g 1g
30003251 250 mg 250mg
30008764 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011271 Supplier Toronto Research Chemicals Supplier No. TRCN4968601G

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 581-89-5
Molecular Formula C10 H7 N O2
Molecular Weight (g/mol) 173.17
Synonym Naphthalene, 2-nitro- (8CI, 9CI, ACI), 2-Nitronaphthalene (ACI), β-Nitronaphthalene
IUPAC Name 2-nitronaphthalene
SMILES [O-][N+](=O)c1ccc2ccccc2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H7NO2/c12-11(13)10-6-5-8-3-1-2-4-9(8)7-10/h1-7H
Formula Weight 173.0477
Chemical Name or Material 2-Nitronaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.