accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHO

CAS RN 582-24-1

IUPAC Name: 2-hydroxy-1-phenylethan-1-one
Synonyms: 2-hydroxyacetophenone glycolophenone
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: ZWVHTXAYIKBMEE-UHFFFAOYSA-N
SMILES: OCC(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)