CAS RN 58409-59-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bucumolol, TRC
CAS: 58409-59-9 Molecular Formula: C17H23NO4 Molecular Weight (g/mol): 305.37 Synonym: 8-[3-[(1,1-Dimethylethyl)amino]-2-hydroxypropoxy]-5-methyl-2H-1-benzopyran-2-one; dl-Bucumolol IUPAC Name: 8-[3-(tert-butylamino)-2-hydroxypropoxy]-5-methylchromen-2-one SMILES: Cc1ccc(OCC(O)CNC(C)(C)C)c2OC(=O)C=Cc12