accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 58452-00-9

H3COOOHO

CAS RN 58452-00-9

IUPAC Name: 3-(benzyloxy)-4-methoxybenzoic acid
Synonyms: 3-(benzyloxy)-4-methoxybenzoic acid
Molecular Weight (g/mol): 258.27
Molecular Formula: C15H14O4
InChi Key: YPDXIGBSOBESNI-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OCC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results