CAS RN 58493-49-5
CAS RN 58493-49-5
IUPAC Name:
(9E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]octadec-9-enamide
Synonyms:
(9E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]octadec-9-enamide
Molecular Weight (g/mol):
417.63
Molecular Formula:
C26H43NO3
InChi Key:
OPZKBPQVWDSATI-ZHACJKMWSA-N
SMILES:
CCCCCCCC\C=C\CCCCCCCC(=O)NCC1=CC=C(O)C(OC)=C1
1
–
1
of
1
results
1