missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Olvanil
GREENER_CHOICE

Product Code. 18084482
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18084482 50 mg 50mg
18084152 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18084482 Supplier Tocris Bioscience™ Supplier No. 0934/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent vanilloid receptor agonist

Potent vanilloid receptor agonist (pEC50 values are 8.1 and 7.7 at rat and human VR1 receptors respectively). Also blocks anandamide uptake (IC50 = 9 μM) and may bind to CB1 cannabinoid receptors. Antinociceptive following systemic administration.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58493-49-5
Molecular Formula C26H43NO3

Specifications

Quantity 50 mg
Formula Weight 417.63
Purity >98%
Molecular Weight (g/mol) 417.634
Chemical Name or Material Olvanil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.