accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OH

CAS RN 587-03-1

IUPAC Name: (3-methylphenyl)methanol
Synonyms: 3-methylbenzyl alcohol M-methylbenzyl alcohol
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: JJCKHVUTVOPLBV-UHFFFAOYSA-N
SMILES: CC1=CC=CC(CO)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results