accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 59-42-7

H3CNH(R)OHHO

CAS RN 59-42-7

IUPAC Name: 3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol
Synonyms: phenylephrine consdrin neosympatol ak-nefrin L-phenylephedrine alconefrin nasal drops 50 alconefrin nasal drops 12 alconefrin nasal drops 25 m-sympathol nostril spray pump mild
Molecular Weight (g/mol): 167.21
Molecular Formula: C9H13NO2
InChi Key: SONNWYBIRXJNDC-VIFPVBQESA-N
SMILES: CNC[C@H](O)C1=CC=CC(O)=C1
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results