missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylephrine, TRC

Product Code. 30137663
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137663 100 mg 100mg
30134129 250 mg 250mg
30008991 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137663 Supplier Toronto Research Chemicals Supplier No. TRCP320688100MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-42-7
Molecular Formula C9 H13 N O2
Molecular Weight (g/mol) 167.2
Synonym Phenylephrine, (1R)-1-(3-Hydroxyphenyl)-2-(methylamino)ethanol, Benzenemethanol, 3-hydroxy-α-[(methylamino)methyl]-, (αR)-, Benzenemethanol, 3-hydroxy-α-[(methylamino)methyl]-, (R)-, Benzyl alcohol, m-hydroxy-α-[(methylamino)methyl]-, (-)- (7CI,8CI), (αR)-3-Hydroxy-α-[(methylamino)methyl]benzenemethanol, (-)-Phenylephrine, (-)-m-Hydroxy-α-(methylaminomethyl)benzyl alcohol, (-)-m-Oxedrine, (-)-m-Synephrine, (R)-(-)-Phenylephrine, (R)-3-[1-Hydroxy-2-(methylamino)ethyl]phenol, (R)-Phenylephrine, 3-[(1R)-1-Hydroxy-2-(methylamino)ethyl]phenol, Iriphrin, L-Phenylephedrine, Mesaton, Mesatone, Metaoxedrin, Metaoxedrine, Metasympatol, Metasynephrine, Mezaton, Neo-Synephrine, R(-)-Mezaton, Visadron, l-Phenylephrine, l-m-Hydroxy-α-[(methylamino)methyl]benzyl alcohol, m-Methylaminoethanolphenol, m-Oxedrine, m-Sympathol, m-Sympatol, m-Synephrine
IUPAC Name 3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol
SMILES CNC[C@H](O)c1cccc(O)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H13NO2/c1-10-6-9(12)7-3-2-4-8(11)5-7/h2-5,9-12H,6H2,1H3/t9-/m0/s1
Formula Weight 167.0946
Chemical Name or Material Phenylephrine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.