accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 590-67-0

CH3HO

CAS RN 590-67-0

IUPAC Name: 1-methylcyclohexan-1-ol
Synonyms: methylcyclohexanol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: VTBOTOBFGSVRMA-UHFFFAOYSA-N
SMILES: CC1(O)CCCCC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)