accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3O

CAS RN 590-86-3

IUPAC Name: 3-methylbutanal
Synonyms: isovaleraldehyde 3-methylbutanal (substance) aldehyde isovalerianique isovalerylaldehyde
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: YGHRJJRRZDOVPD-UHFFFAOYSA-N
SMILES: CC(C)CC=O
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results