accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3HO(R)(R)HOO(R)OOCH3(E)

CAS RN 59122-46-2

IUPAC Name: methyl 7-[(1R,2R,3R)-3-hydroxy-2-[(1E)-4-hydroxy-4-methyloct-1-en-1-yl]-5-oxocyclopentyl]heptanoate
Synonyms: misoprostol apo misoprostol grunenthal brand of misoprostol glefos misoprostol (substance) searle brand of misoprostol misoprostol searle brand misoprostol grunenthal brand (11α,13E)-isomer misoprostol novo misoprostol
Molecular Weight (g/mol): 382.54
Molecular Formula: C22H38O5
InChi Key: OJLOPKGSLYJEMD-URPKTTJQSA-N
SMILES: CCCCC(C)(O)C\C=C\[C@H]1[C@H](O)CC(=O)[C@@H]1CCCCCCC(=O)OC
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)
Product Line 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results