accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 592-20-1

IUPAC Name: 2-oxopropyl acetate
Synonyms: 2-propanone, 1-(acetyloxy)-
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: DBERHVIZRVGDFO-UHFFFAOYSA-N
SMILES: CC(=O)COC(C)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results