accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 593-56-6

IUPAC Name: hydrogen O-methylhydroxylamine chloride
Synonyms: methoxyamine
Molecular Weight (g/mol): 83.52
Molecular Formula: CH6ClNO
InChi Key: XNXVOSBNFZWHBV-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CON
Molecular Weight (g/mol) 
  • (7)
  • (6)
Quantity 
  • (2)
  • (2)
  • (5)
  • (4)
Physical Form 
  • (6)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (3)
  • (7)
  • (6)
  • (2)
  • (2)
  • (5)
  • (4)
  • (6)
  • (2)
  • (3)