missing translation for 'onlineSavingsMsg'
Learn More

Methoxylamine Hydrochloride, TRC

Product Code. 30470805
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30470805 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30470805 Supplier Toronto Research Chemicals Supplier No. TRCM2627201G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methoxylamine Hydrochloride
CAS 593-56-6
Molecular Formula C H5 N O . Cl H
Quantity 1 g
Synonym Hydroxylamine, O-methyl-, hydrochloride (1:1) (ACI), Hydroxylamine, O-methyl-, hydrochloride (8CI, 9CI), (Aminooxy)methane hydrochloride, Hydroxylamine methyl ether hydrochloride, Methoxyamine hydrochloride, Methoxyammonium chloride, Methoxylamine hydrochloride, N-Methoxylamine hydrochloride, O-Methoxyamine hydrochloride, O-Methylhydroxyamine hydrochloride, O-Methylhydroxylamine hydrochloride
InChI Formula InChI=1S/CH5NO.ClH/c1-3-2;/h2H2,1H3;1H
SMILES Cl.CON
IUPAC Name O-methylhydroxylamine;hydrochloride
Molecular Weight (g/mol) 83.52
Formula Weight 83.0138
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.