accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5932-53-6

CH3NHOO

CAS RN 5932-53-6

IUPAC Name: 6-hydroxy-8-methyl-8-azabicyclo[3.2.1]octan-3-one
Synonyms: 6-hydroxy-8-methyl-8-azabicyclo[3.2.1]octan-3-one
Molecular Weight (g/mol): 155.2
Molecular Formula: C8H13NO2
InChi Key: UOHSTKWPZWFYTF-UHFFFAOYNA-N
SMILES: CN1C2CC(O)C1CC(=O)C2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)