accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3COHOHO

CAS RN 594-61-6

IUPAC Name: 2-hydroxy-2-methylpropanoic acid
Synonyms: acetonate (ch3)2cohcooh HIBA acetonic acid hydroxyisobutyrate α-hydroxy-I-butyric acid α-HIB α-hib
Molecular Weight (g/mol): 104.11
Molecular Formula: C4H8O3
InChi Key: BWLBGMIXKSTLSX-UHFFFAOYSA-N
SMILES: CC(C)(O)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)