accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 59741-04-7

IUPAC Name: 2-isocyanato-1-methoxy-4-methylbenzene
Synonyms: 2-isocyanato-1-methoxy-4-methylbenzene
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: IDOHLSFNBKNRRJ-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C)C=C1)N=C=O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results