accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5977-14-0

IUPAC Name: 3-oxobutanamide
Synonyms: acetoacetamide AAA
Molecular Weight (g/mol): 101.11
Molecular Formula: C4H7NO2
InChi Key: GCPWJFKTWGFEHH-UHFFFAOYSA-N
SMILES: CC(=O)CC(N)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Industry Type 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results