accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 599-00-8

OOFFF2H

CAS RN 599-00-8

IUPAC Name: trifluoroethan(²H)oic acid
Synonyms: trifluoroethan(²H)oic acid
Molecular Weight (g/mol): 115.03
Molecular Formula: C2HF3O2
InChi Key: DTQVDTLACAAQTR-DYCDLGHISA-N
SMILES: [2H]OC(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (10)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (9)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (7)
  • (2)
  • (2)
  • (10)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (9)
  • (2)
  • (2)