accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClNH2OH

CAS RN 60-19-5

IUPAC Name: hydrogen 4-(2-aminoethyl)phenol chloride
Synonyms: hydrogen tyramine chloride
Molecular Weight (g/mol): 173.64
Molecular Formula: C8H12ClNO
InChi Key: RNISDHSYKZAWOK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCC1=CC=C(O)C=C1
Quantity 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results