missing translation for 'onlineSavingsMsg'
Learn More

Tyramine Hydrochloride, TRC

Product Code. 17962895
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962895 100 g 100g
17953675 5 g 5g
17913675 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17962895 Supplier Toronto Research Chemicals Supplier No. TRCT898493100G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-19-5
Molecular Formula C8 H11 N O . Cl H
Molecular Weight (g/mol) 173.64
Synonym Phenol, 4-(2-aminoethyl)-, hydrochloride (1:1), Phenol, 4-(2-aminoethyl)-, hydrochloride (9CI), Phenol, p-(2-aminoethyl)-, hydrochloride (8CI), 2-(4-Hydroxyphenyl)ethylamine hydrochloride, 4-Hydroxy-β-phenethylamine hydrochloride, 4-Hydroxyphenethylamine hydrochloride, 4-Hydroxyphenylethylamine hydrochloride, Mydrial, Tyramine hydrochloride, Tyrosam, Uteramin, p-Tyramine hydrochloride
IUPAC Name 4-(2-aminoethyl)phenol;hydrochloride
SMILES Cl.NCCc1ccc(O)cc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H11NO.ClH/c9-6-5-7-1-3-8(10)4-2-7;/h1-4,10H,5-6,9H2;1H
Formula Weight 173.0607
Chemical Name or Material Tyramine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.