CAS RN 6001-64-5
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, 98%
CAS: 6001-64-5 Molecular Formula: C4H11Cl3O2 Molecular Weight (g/mol): 197.48 MDL Number: MFCD00004461 InChI Key: HBARVHNVVKZUGS-UHFFFAOYSA-N Synonym: c4h7cl3o.1/2h2o,1,1,1-trichloro-2-methyl-2-propanol hemihydrate,chlorobutanol hydrogen ion hydrate PubChem CID: 102594540 IUPAC Name: molecular hydrogen;1,1,1-trichloro-2-methylpropan-2-ol;hydrate SMILES: [HH].CC(C)(C(Cl)(Cl)Cl)O.O
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, 98%
CAS: 6001-64-5 Molecular Formula: 0·5 H2O Molecular Weight (g/mol): 186.47 MDL Number: MFCD02179352 InChI Key: HBARVHNVVKZUGS-UHFFFAOYSA-N Synonym: c4h7cl3o.1/2h2o,1,1,1-trichloro-2-methyl-2-propanol hemihydrate,chlorobutanol hydrogen ion hydrate PubChem CID: 102594540 IUPAC Name: molecular hydrogen;1,1,1-trichloro-2-methylpropan-2-ol;hydrate SMILES: [HH].CC(C)(C(Cl)(Cl)Cl)O.O
Chlorobutanol Hemihydrate, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Chlorbutanol Hemihydrate, TRC
CAS: 6001-64-5 Molecular Formula: 2 C4 H7 Cl3 O . H2 O Molecular Weight (g/mol): 372.93 Synonym: 2-Propanol, 1,1,1-trichloro-2-methyl-, hemihydrate (8CI),1,1,1-Trichloro-2-methyl-2-propanol hemihydrate,Acetonchloroform hemihydrate IUPAC Name: 1,1,1-trichloro-2-methylpropan-2-ol;hydrate SMILES: O.CC(C)(O)C(Cl)(Cl)Cl.CC(C)(O)C(Cl)(Cl)Cl
Chlorobutanol Hemihydrate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.