accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3NCH3H3CNCH3CH3

CAS RN 603-48-5

IUPAC Name: 4-{bis[4-(dimethylamino)phenyl]methyl}-N,N-dimethylaniline
Synonyms: 4-{bis[4-(dimethylamino)phenyl]methyl}-N,N-dimethylaniline
Molecular Weight (g/mol): 373.54
Molecular Formula: C25H31N3
InChi Key: OAZWDJGLIYNYMU-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Color 
  • (2)
  • (2)
Grade 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)