missing translation for 'onlineSavingsMsg'
Learn More

Leuco Gentian Violet, TRC

Product Code. 30138666
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138666 1 g 1g
30136401 5 g 5g
17994585 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138666 Supplier Toronto Research Chemicals Supplier No. TRCL3302001G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-48-5
Molecular Formula C25 H31 N3
Molecular Weight (g/mol) 373.53
Synonym Colorless crystal violet, Reduced crystal violet
IUPAC Name 4-[bis[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
SMILES CN(C)c1ccc(cc1)C(c2ccc(cc2)N(C)C)c3ccc(cc3)N(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C25H31N3/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6/h7-18,25H,1-6H3
Formula Weight 373.2518
Chemical Name or Material Leucocrystal Violet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.