CAS RN 603122-40-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Methoxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester, 95%
CAS: 603122-40-3 Molecular Formula: C15H21BO5 Molecular Weight (g/mol): 292.14