accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CO

CAS RN 6048-82-4

IUPAC Name: 1-phenyldecan-1-one
Synonyms: 1-phenyldecan-1-one
Molecular Weight (g/mol): 232.37
Molecular Formula: C16H24O
InChi Key: QQXJNLYVPPBERR-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)